Learn More
Waters Corp Amino Acid Standard, Hydrolysate, 10 x 1 mL, 2.5 mM
Catalog No. 50785966 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This amino acid standard is a mixture designed for identification and quantitative amino acid analysis. It includes 18 amino acids, with 17 hydrolysate amino acids each at 2.5 mM and cystine at 1.25 mM, supplied in 10 x 1 mL ampules.
  • Contains 17 hydrolysate amino acids at 2.5 mM and cystine at 1.25 mM.
  • Supplied in 10 x 1 mL ampules.
  • Designed for identification and quantitative amino acid analysis.
Catalog No. 50-785-966 Supplier Waters Corp Supplier No. WAT088122
This amino acid standard is a mixture designed for identification and quantitative amino acid analysis. It includes 18 amino acids, with 17 hydrolysate amino acids each at 2.5 mM and cystine at 1.25 mM, supplied in 10 x 1 mL ampules.
  • Contains 17 hydrolysate amino acids at 2.5 mM and cystine at 1.25 mM.
  • Supplied in 10 x 1 mL ampules.
  • Designed for identification and quantitative amino acid analysis.