Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amino Acid Standards, For Protein Hydrolysates, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111004480 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4480 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4480 Supplier MilliporeSigma Supelco Supplier No. A97815ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino acids listed are 0.5 mumole/mL except L-cystine at 0.25 mumole/mL in 0.2 N sodium citrate, pH 2.2.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Quantity 5 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.