Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amino Acids Large Model Set
SCI_ED

Catalog No. S94026
Change view
Click to view available options
Amino Acids large model set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S94026 Amino Acids large model set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94026 Supplier Ryler Enterprises Supplier No. AA2

missing translation for 'validMainframeMsg'

Includes

Kit Contents:

  • 52 white 1-peg hydrogen atom centers
  • 2 white 2-peg hydrogen atom centers
  • 2 blue 4-peg nitrogen atom centers
  • 9 blue 3-peg nitrogen atom centers
  • 14 red 2-peg oxygen atom centers
  • 2 red 3-peg oxygen atom centers
  • 3 yellow 2-peg sulfur atom centers
  • 22 black 4-peg carbon atom centers
  • 7 black 3-peg carbon atom centers
  • 90-1.25 in. PVC, clear bonds (single bonds)
  • 14-1.625 in. PVC, clear bonds (double bonds)
  • 3-2 in. PVC, white bonds (H bonds)
  • Lab procedure
  • Answer key

Build five amino acids simultaneously with this kit.

An expanded of Amino Acids Small Model Set (S94025). Students can build five amino acids simultaneously with this kit to investigate D and L forms of amino acids, hydrogen bonding, dehydration synthesis, peptide formation, the meaning of alpha amino acid, primary and secondary structure of proteins. As separate investigations, students can make models of each of the common 20 alpha amino acids.

  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing

Specifications

For Use With (Application) To investigate D and L forms of Amino Acids, Hydrogen bonding, dehydration synthesis
Includes 52 white 1-peg Hydrogen atom centers, 2 white 2-peg Hydrogen atom centers, 2 blue 4-peg Nitrogen atom centers, 9 blue 3-peg Nitrogen atom centers, 14 red 2-peg Oxygen atom centers, 2 red 3-peg Oxygen atom centers, 3 yellow 2-peg sulfur atom centers, 22 black 4-peg Carbon atom centers, 7 black 3-peg Carbon atom centers, Ninety 1.25 in. PVC, clear bonds (single bonds), Fourteen 1.625 in. PVC, clear bonds (double bonds), 3 to 2 in. PVC, white bonds (H bonds), lab procedure, answer key
Size Large
Product Type Model Set, Amino Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.