Learn More
FUJIFILM Biosciences Amino Acids Mixture Standard
Catalog No. 5000450338
Encompass_Preferred
This product contains ammonium chloride generated by the hydrolysis of 17 amino acids glutamine and asparagine It can be used for amino acid composition analysis and quantitative analysis This product uses either analytical reference materials with a guarantee of amino acid impurities or reference materials with a guarantee of SI traceability as raw materials A lineup of three products with different compositions
Catalog No. 50-004-50338 Supplier FUJIFILM Biosciences Supplier No. 01928151
This product contains ammonium chloride generated by the hydrolysis of 17 amino acids glutamine and asparagine It can be used for amino acid composition analysis and quantitative analysis This product uses either analytical reference materials with a guarantee of amino acid impurities or reference materials with a guarantee of SI traceability as raw materials A lineup of three products with different compositions