Learn More
FUJIFILM Biosciences Amino Acids Mixture StND Sol
Catalog No. 5000450387
Encompass_Preferred
This product is a mixture standard solution containing 12 amino acids including 3 basic amino acids It can be used for amino acid composition analysis and quantitative analysis This product uses either analytical reference materials with a guarantee of amino acid impurities or reference materials with a guarantee of SI traceability as raw materials A lineup of three products with different compositions
Catalog No. 50-004-50387 Supplier FUJIFILM Biosciences Supplier No. 01628144
This product is a mixture standard solution containing 12 amino acids including 3 basic amino acids It can be used for amino acid composition analysis and quantitative analysis This product uses either analytical reference materials with a guarantee of amino acid impurities or reference materials with a guarantee of SI traceability as raw materials A lineup of three products with different compositions