Learn More
Bioworld AMINO COUPLING KIT
Supplier:  Bioworld 201500001
Encompass_Preferred
bio-World's Amino CouplingKitconsists of aldehyde terminated Separopore(Agarose)(crosslinked 4% agarose beads) that has been activated to covalently immobilize antibodies and other proteins through primary amine groups. Once a protein is...
Catalog No. 50-197-556