Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amino-Functionalized Silica Gel, Spherical, 40-75 μm

Catalog No. 111012885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2885 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2885 Supplier MilliporeSigma™ Supelco™ Supplier No. 792971KG
Add to Cart
Add to Cart

Amino-functionalized silica gel spherical is monodisperse in size and shape.

Amino-functionalized silica gel spherical is prepared and characterized by thermogravimetery, X-ray diffraction, FT-IR, scanning electron microscopy (SEM) and N2 adsorption-desorption techniques.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 40 to 75 μm
Pore Size 110 Å
Grade High-purity Grade
Quantity 1 kg
Surface Area (Metric) 500 m2/g (approx.)
Description Nitrogen content, >1.0%
Physical Form Powder
CAS 126850-14-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.