Learn More
Abcam 7-Amino-4-methylcoumarin, Fluorescent labeling reagent, 100MG
Catalog No. 50996201
Encompass_Preferred
MW 175.18 Da, Purity >99%. Fluorescent probe to analyse glycoproteins' monosaccharides and N-linked oligosaccharides. Antibacterial and antifungal agent in vitro. Used for spectrophotometric determination of small amounts of nitrite.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-201 Supplier Abcam Supplier No. AB142121100MG
MW 175.18 Da, Purity >99%. Fluorescent probe to analyse glycoproteins' monosaccharides and N-linked oligosaccharides. Antibacterial and antifungal agent in vitro. Used for spectrophotometric determination of small amounts of nitrite.
The product is subject to the following: Abcam Restricted Use Statement