Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ε-Amino-N-caproic Acid, 99.8%, MP Biomedicals™

Catalog No. ICN10164591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10164591 1 kg
ICN10164525 25 g
ICN10164580 100 g
ICN10164590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10164591 Supplier MP Biomedicals, Inc Supplier No. 0210164591
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN

Specifications

Color White
Density 1.48g/mL
Assay Percent Range 99.8%
Quantity 1 kg
Formula Weight 131.2
Physical Form Crystalline Powder
Chemical Name or Material ε-Amino-N-caproic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.