Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ε-Amino-N-Caproic Acid, MP Biomedicals™

Catalog No. MP210164594 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210164594 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210164594 Supplier MP Biomedicals, Inc Supplier No. 0210164594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits plasmin and carboxypeptidase B. ε-Amino-N-Caproic Acid, MP Biomedicals is a lysine analog that promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. It is has been utilized i

EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN

Specifications

Melting Point 205°C to 215°C
Density 1.48g/mL (lit.)
Quantity 5 kg
Solubility Information Soluble in water (50mg/mL); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.
Formula Weight 131.2
Physical Form Powder
Recommended Storage Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.