Learn More
Waters Corp Amino (NH2) Column, 125 Å, 10 μm Particle, 3.9 mm ID
Catalog No. 502392323 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Carbohydrate Analysis Column with covalently bonded amino packing is ideal for separating low molecular weight sugars (mono-, di-, and trisaccharides). It typically uses a mobile phase of 65-85% acetonitrile in water and operates at ambient to 35°C.
  • Covalently bonded amino packing.
  • Separates low molecular weight sugars (mono-, di-, trisaccharides).
  • Uses 65-85% acetonitrile in water mobile phase.
  • Operates from ambient to 35°C.
  • Equipped with an electronic eConnect column tag.
  • eConnect tag provides unique, column-specific information for tracking.
  • Compatible with any HPLC system.
  • Optimized for Alliance iS System with Empower software.
Catalog No. 50-239-2323 Supplier Waters Corp Supplier No. WAT084038RF
This Carbohydrate Analysis Column with covalently bonded amino packing is ideal for separating low molecular weight sugars (mono-, di-, and trisaccharides). It typically uses a mobile phase of 65-85% acetonitrile in water and operates at ambient to 35°C.
  • Covalently bonded amino packing.
  • Separates low molecular weight sugars (mono-, di-, trisaccharides).
  • Uses 65-85% acetonitrile in water mobile phase.
  • Operates from ambient to 35°C.
  • Equipped with an electronic eConnect column tag.
  • eConnect tag provides unique, column-specific information for tracking.
  • Compatible with any HPLC system.
  • Optimized for Alliance iS System with Empower software.