Learn More
Medchemexpress LLC Amino-PEG1-C2-acid | 144942-89-2 | 98.0% | 133.15 | 1 G
SDP
Catalog No. 5000349842
Encompass_Preferred
Amino-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This linker is utilized in creating compounds that facilitate the degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates degradation of target proteins
  • For research use only
Catalog No. 50-003-49842 Supplier Medchemexpress LLC Supplier No. HY1400021G
Amino-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This linker is utilized in creating compounds that facilitate the degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates degradation of target proteins
  • For research use only