Learn More
Medchemexpress LLC Amino-PEG1-C2-acid | 144942-89-2 | 98.0% | 133.15 | 100 MG
SDP
Catalog No. 5000350742
Encompass_Preferred
Amino-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This product is for research use only. PROTACs function by containing two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein. They harness the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Suitable for research use only
Catalog No. 50-003-50742 Supplier Medchemexpress LLC Supplier No. HY140002100MG
Amino-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This product is for research use only. PROTACs function by containing two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein. They harness the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Suitable for research use only