Learn More
Selleck Chemical LLC Amino-PEG1-C2-acid S1872-1g
SDP
Catalog No. 503109291
Encompass_Preferred
Amino-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs
Catalog No. 50-310-9291 Supplier Selleck Chemical LLC Supplier No. S18721G
Amino-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs