Learn More
Medchemexpress LLC Amino-PEG10-acid | 2170987-85-4 | 529.62 | 2 MG
SDP
Catalog No. 5000349209
Encompass_Preferred
Amino-PEG10-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that link an E3 ubiquitin ligase ligand with a target protein ligand, enabling the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Chemical formula: C23H47NO12
  • Suitable for research use only
  • Shipping at room temperature in continental US
  • Store product under recommended conditions in the Certificate of Analysis
Catalog No. 50-003-49209 Supplier Medchemexpress LLC Supplier No. HY1401762MG
Amino-PEG10-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that link an E3 ubiquitin ligase ligand with a target protein ligand, enabling the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Chemical formula: C23H47NO12
  • Suitable for research use only
  • Shipping at room temperature in continental US
  • Store product under recommended conditions in the Certificate of Analysis