Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG11-amine | 479200-82-3 | 95.0% | 100 MG
SDP

Catalog No. 5000338305
Encompass_Preferred

Amino-PEG11-amine is a PEG-based PROTAC linker designed to combine mono diethylstilbestrol (DES)-based ligands. This offers an alternative approach for creating more selective and active ER antagonists, useful in the endocrine treatment of breast cancer. The product appears as an oil with a white to off-white color. Its molecular weight is 544.68 and its formula is C24H52N2O11. This product is for research use only and not for sale to patients.

  • PEG-based PROTAC linker
  • Combines mono diethylstilbestrol (DES)-based ligands
  • Supports development of selective ER antagonists
  • Potential for breast cancer endocrine therapy
  • Appears as an oil
  • Color: white to off-white
  • Molecular weight: 544.68
  • Chemical formula: C24H52N2O11
  • For research use only

Catalog No. 50-003-38305 Supplier Medchemexpress LLC Supplier No. HY130411100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG11-amine is a PEG-based PROTAC linker designed to combine mono diethylstilbestrol (DES)-based ligands. This offers an alternative approach for creating more selective and active ER antagonists, useful in the endocrine treatment of breast cancer. The product appears as an oil with a white to off-white color. Its molecular weight is 544.68 and its formula is C24H52N2O11. This product is for research use only and not for sale to patients.

  • PEG-based PROTAC linker
  • Combines mono diethylstilbestrol (DES)-based ligands
  • Supports development of selective ER antagonists
  • Potential for breast cancer endocrine therapy
  • Appears as an oil
  • Color: white to off-white
  • Molecular weight: 544.68
  • Chemical formula: C24H52N2O11
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.