Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG11-amine | 479200-82-3 | 95.0% | 50 MG
SDP

Catalog No. 5000338441
Encompass_Preferred

Amino-PEG11-amine is a PEG-based (12 units) PROTAC linker used to combine two mono diethylstilbestrol (DES)-based ligands. It offers an alternative strategy for creating more selective and active ER antagonists for the endocrine therapy of breast cancer. It has a molecular weight of 544.68 and a chemical formula of C24H52N2O11.

  • Purity: 95.0%
  • Appearance: Oil
  • Color: White to off-white
  • For research use only.

Catalog No. 50-003-38441 Supplier Medchemexpress LLC Supplier No. HY13041150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG11-amine is a PEG-based (12 units) PROTAC linker used to combine two mono diethylstilbestrol (DES)-based ligands. It offers an alternative strategy for creating more selective and active ER antagonists for the endocrine therapy of breast cancer. It has a molecular weight of 544.68 and a chemical formula of C24H52N2O11.

  • Purity: 95.0%
  • Appearance: Oil
  • Color: White to off-white
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.