Learn More
TARGETMOL CHEMICALS INC Amino-PEG12-acid 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T17405100MG
Encompass_Preferred
Also available in 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Amino-PEG12-acid, a PEG-based PROTAC linker, is utilized in PROTAC synthesis. Purity 98.61%
Catalog No. 50-287-8544