Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG12-amine | 361543-12-6 | 99.9% | 588.73 | 1 G
SDP

Catalog No. 5000365322
Encompass_Preferred

Amino-PEG12-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is intended for research use only.

  • Used in the synthesis of PROTACs
  • Molecular weight of 588.73
  • Purity of 99.92% by HPLC
  • Colorless to off-white solid-liquid mixture
  • Store at 4°C, protected from light

Catalog No. 50-003-65322 Supplier Medchemexpress LLC Supplier No. 1GHY133327
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG12-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is intended for research use only.

  • Used in the synthesis of PROTACs
  • Molecular weight of 588.73
  • Purity of 99.92% by HPLC
  • Colorless to off-white solid-liquid mixture
  • Store at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.