Learn More
Medchemexpress LLC Amino-PEG12-amine | 361543-12-6 | HY-133327 | 99.9% | 588.73 | 250 MG
SDP
Catalog No. 5000365668
Encompass_Preferred
Amino-PEG12-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules with two different ligands connected by a linker: one for an E3 ubiquitin ligase and another for a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Appears as a colorless to off-white solid-liquid mixture
  • Has a purity of 99.92%
Catalog No. 50-003-65668 Supplier Medchemexpress LLC Supplier No. 250MGHY133327
Amino-PEG12-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules with two different ligands connected by a linker: one for an E3 ubiquitin ligase and another for a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Appears as a colorless to off-white solid-liquid mixture
  • Has a purity of 99.92%