Learn More
Medchemexpress LLC Amino-PEG12-Boc | 1383814-00-3 | 98.0% | C31H63NO14 | 100 MG
SDP
Catalog No. 5000353018
Encompass_Preferred
Amino-PEG12-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-53018 Supplier Medchemexpress LLC Supplier No. HY140197100MG
Amino-PEG12-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system to degrade target proteins