Learn More
Medchemexpress LLC Amino-PEG12-Boc | 1383814-00-3 | 98.0% | C31H63NO14 | 25 MG
SDP
Catalog No. 5000352653
Encompass_Preferred
Amino-PEG12-Boc is a PEG-based PROTAC linker for synthesizing PROTACs. PROTACs (Proteolysis Targeting Chimeras) are designed with two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein, to exploit the ubiquitin-proteasome system for selective degradation of proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-52653 Supplier Medchemexpress LLC Supplier No. 25MGHY140197
Amino-PEG12-Boc is a PEG-based PROTAC linker for synthesizing PROTACs. PROTACs (Proteolysis Targeting Chimeras) are designed with two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein, to exploit the ubiquitin-proteasome system for selective degradation of proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins