Learn More
Medchemexpress LLC Amino-PEG16-acid | 2922656-52-6 | 95.0% | C35H71NO18 | 50 MG
SDP
Catalog No. 5000357194
Encompass_Preferred
Amino-PEG16-acid is a PEG-based PROTAC linker. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-57194 Supplier Medchemexpress LLC Supplier No. HY14017950MG
Amino-PEG16-acid is a PEG-based PROTAC linker. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.