Learn More
Medchemexpress LLC Amino-peg2-c2-acid | 791028-27-8 | 99.7% | 177.20 g/mol | C7H15NO4 | 1 G
SDP
Catalog No. 5000202851
Encompass_Preferred
Amino-PEG2-C2-acid is a bifunctional PEG-based linker (molecular formula C7H15NO4, molecular weight 177.20 g/mol) that contains a primary amine and a terminal carboxylic acid. It is used as a cleavable linker in antibody-drug conjugates (ADCs) and as a PROTAC linker for bioconjugation and linker discovery.
  • Contains a primary amine and a terminal carboxylic acid.
  • Cleavable three-unit PEG spacer providing a short, flexible linkage.
  • Applicable for ADC synthesis, PROTAC construction, and general bioconjugation.
  • Supplied with high purity and RP-HPLC documentation.
Catalog No. 50-002-02851 Supplier Medchemexpress LLC Supplier No. HYW0401681G
Amino-PEG2-C2-acid is a bifunctional PEG-based linker (molecular formula C7H15NO4, molecular weight 177.20 g/mol) that contains a primary amine and a terminal carboxylic acid. It is used as a cleavable linker in antibody-drug conjugates (ADCs) and as a PROTAC linker for bioconjugation and linker discovery.
  • Contains a primary amine and a terminal carboxylic acid.
  • Cleavable three-unit PEG spacer providing a short, flexible linkage.
  • Applicable for ADC synthesis, PROTAC construction, and general bioconjugation.
  • Supplied with high purity and RP-HPLC documentation.