Learn More
Medchemexpress LLC Amino-PEG2-(CH2)3COOH | 1263046-77-0 | ≥97.0% | C8H17NO4 | 100 MG
SDP
Catalog No. 5000356762
Encompass_Preferred
Amino-PEG2-(CH2)3COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only. The product appears as a viscous liquid, light yellow to yellow in color.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a viscous liquid
  • Light yellow to yellow in color
Catalog No. 50-003-56762 Supplier Medchemexpress LLC Supplier No. HY140190100MG
Amino-PEG2-(CH2)3COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only. The product appears as a viscous liquid, light yellow to yellow in color.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a viscous liquid
  • Light yellow to yellow in color