Learn More
Medchemexpress LLC Amino-PEG2-CH2CH2-SH (hydrochloride) | 3037676-58-4 | 95.0% | 201.71 g/mol | C6H16ClNO2S | 5 MG
SDP
Catalog No. 5000262826
Encompass_Preferred
Amino-PEG2-CH2CH2-SH (hydrochloride) is a PEG-based bifunctional linker for synthesis of PROTACs and conjugates. It features a primary amino group and a terminal thiol separated by a two-unit PEG spacer, supplied as the hydrochloride salt to aid stability and handling.
  • Provides primary amino and terminal thiol functional groups for conjugation
  • Two-unit PEG spacer offers flexibility and solubility
  • Hydrochloride salt form improves stability and handling
  • High purity suitable for synthetic applications
  • Available in small milligram pack sizes for research use
Catalog No. 50-002-62826 Supplier Medchemexpress LLC Supplier No. HY1321205MG
Amino-PEG2-CH2CH2-SH (hydrochloride) is a PEG-based bifunctional linker for synthesis of PROTACs and conjugates. It features a primary amino group and a terminal thiol separated by a two-unit PEG spacer, supplied as the hydrochloride salt to aid stability and handling.
  • Provides primary amino and terminal thiol functional groups for conjugation
  • Two-unit PEG spacer offers flexibility and solubility
  • Hydrochloride salt form improves stability and handling
  • High purity suitable for synthetic applications
  • Available in small milligram pack sizes for research use