Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG2-NH-Boc | 127828-22-2 | 204.27 | 5 G
SDP

Catalog No. 5000447464
Encompass_Preferred

Amino-PEG2-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, enabling them to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity: 98.0%
  • Molecular weight: 204.27
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-47464 Supplier Medchemexpress LLC Supplier No. HYW0082795G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG2-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, enabling them to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity: 98.0%
  • Molecular weight: 204.27
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.