Learn More
Medchemexpress LLC Amino-PEG24-Boc | 2170987-96-7 | 1012664 | 99.8% | C55H111NO26 | 100 MG
SDP
Catalog No. 5000353486
Encompass_Preferred
Amino-PEG24-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53486 Supplier Medchemexpress LLC Supplier No. HY140199100MG
Amino-PEG24-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins