Learn More
Medchemexpress LLC Amino-peg3-c2-acid | 784105-33-5 | MFCD21607085 | 99.9% | 221.25 g·mol⁻¹ | C9H19NO5 | 1 G
SDP
Catalog No. 5000202854
Encompass_Preferred
Amino-PEG3-C2-acid is a bifunctional PEG-based linker with terminal amino and carboxylic acid groups, designed for conjugation chemistry in PROTAC and antibody-drug conjugate synthesis. Intended for research use only.
  • Short PEG spacer with three ethylene glycol units.
  • Terminal amino and carboxylic acid functional groups.
  • Suitable for PROTAC and ADC linker synthesis.
  • High purity (99.94%).
  • Available in pack sizes including 1 g.
Catalog No. 50-002-02854 Supplier Medchemexpress LLC Supplier No. HYW0401651G
Amino-PEG3-C2-acid is a bifunctional PEG-based linker with terminal amino and carboxylic acid groups, designed for conjugation chemistry in PROTAC and antibody-drug conjugate synthesis. Intended for research use only.
  • Short PEG spacer with three ethylene glycol units.
  • Terminal amino and carboxylic acid functional groups.
  • Suitable for PROTAC and ADC linker synthesis.
  • High purity (99.94%).
  • Available in pack sizes including 1 g.