Learn More
Medchemexpress LLC Amino-PEG3-C2-amine | 929-75-9 | MFCD00628164 | 99.1% | 192.26 g/mol | C8H20N2O3 | 1 G
SDP
Catalog No. 5000303159
Encompass_Preferred
Amino-PEG3-C2-amine is a short polyethylene glycol (PEG) spacer bearing a terminal primary amine, intended for use as a PROTAC linker and general bioconjugation. It provides three ethylene glycol units to improve solubility and flexibility, while the amino group enables straightforward coupling chemistry for building complex conjugates.
  • Short PEG spacer (three ethylene glycol units) for flexibility and solubility.
  • Terminal primary amine suitable for coupling to carboxylic acids and activated esters.
  • Liquid form simplifies handling and accurate dosing.
  • High purity supports reliable synthetic outcomes.
  • Useful in PROTAC linker synthesis and general bioconjugation applications.
Catalog No. 50-003-03159 Supplier Medchemexpress LLC Supplier No. HYW0150881G
Amino-PEG3-C2-amine is a short polyethylene glycol (PEG) spacer bearing a terminal primary amine, intended for use as a PROTAC linker and general bioconjugation. It provides three ethylene glycol units to improve solubility and flexibility, while the amino group enables straightforward coupling chemistry for building complex conjugates.
  • Short PEG spacer (three ethylene glycol units) for flexibility and solubility.
  • Terminal primary amine suitable for coupling to carboxylic acids and activated esters.
  • Liquid form simplifies handling and accurate dosing.
  • High purity supports reliable synthetic outcomes.
  • Useful in PROTAC linker synthesis and general bioconjugation applications.