Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG3-C2-azido | 134179-38-7 | MFCD00269874 | 99.7% | 218.25 g/mol | C8H18N4O3 | 500 MG
SDP

Catalog No. 5000301995
Encompass_Preferred

Amino-PEG3-C2-azido is a PEG-based PROTAC linker and click-chemistry reagent containing an azide functional group. It enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition for conjugation of azide- and alkyne-bearing fragments, making it suitable for linker assembly in degrader synthesis and bioconjugation workflows.

  • High purity (99.67%) suitable for synthetic applications.
  • Azide functionality enables CuAAC and SPAAC conjugations.
  • Compatible with assembly of PROTAC linkers and bioconjugates.
  • Molecular weight 218.25 g/mol; formula C8H18N4O3.
  • Available in multiple pack sizes to support small- and larger-scale work.
  • Store at -20°C, protect from light, under inert gas for stability.

Catalog No. 50-003-01995 Supplier Medchemexpress LLC Supplier No. HYW021401500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG3-C2-azido is a PEG-based PROTAC linker and click-chemistry reagent containing an azide functional group. It enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition for conjugation of azide- and alkyne-bearing fragments, making it suitable for linker assembly in degrader synthesis and bioconjugation workflows.

  • High purity (99.67%) suitable for synthetic applications.
  • Azide functionality enables CuAAC and SPAAC conjugations.
  • Compatible with assembly of PROTAC linkers and bioconjugates.
  • Molecular weight 218.25 g/mol; formula C8H18N4O3.
  • Available in multiple pack sizes to support small- and larger-scale work.
  • Store at -20°C, protect from light, under inert gas for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.