Learn More
Medchemexpress LLC Amino-PEG3-C2-sulfonic acid | 1817735-43-5 | 99.47% | 257.30 | 100 MG
SDP
Catalog No. 5000350271
Encompass_Preferred
Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Contains two distinct ligands connected by a linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50271 Supplier Medchemexpress LLC Supplier No. HY140001100MG
Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Contains two distinct ligands connected by a linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins