Learn More
Medchemexpress LLC Amino-PEG3-C2-sulfonic acid | 1817735-43-5 | 99.5% | 257.30 | 50 MG
SDP
Catalog No. 5000350272
Encompass_Preferred
Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules containing two ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein. Their application involves exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Applicable in cancer and cancer targeted therapy
Catalog No. 50-003-50272 Supplier Medchemexpress LLC Supplier No. HY14000150MG
Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules containing two ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein. Their application involves exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Applicable in cancer and cancer targeted therapy