Learn More
Medchemexpress LLC Amino-PEG3-CH2COOH | 134978-99-7 | 99.9% | C8H17NO5 | 100 MG
SDP
Catalog No. 5000353074
Encompass_Preferred
Amino-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53074 Supplier Medchemexpress LLC Supplier No. HY140189100MG
Amino-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins