Learn More
Medchemexpress LLC Amino-peg3-ch2cooh | 134978-99-7 | 99.9% | C8H17NO5 | 250 MG
SDP
Catalog No. 5000352827
Encompass_Preferred
Amino-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Has a chemical formula of C8H17NO5.
  • Features a molecular weight of 207.22.
  • Offers a purity of 99.9%.
  • Requires storage at 4°C, protected from light.
  • For solutions, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.
Catalog No. 50-003-52827 Supplier Medchemexpress LLC Supplier No. 250MGHY140189
Add to Cart
Add to Cart
Amino-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Has a chemical formula of C8H17NO5.
  • Features a molecular weight of 207.22.
  • Offers a purity of 99.9%.
  • Requires storage at 4°C, protected from light.
  • For solutions, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.