Learn More
Medchemexpress LLC HY-W008005 100mg Medchemexpress, Amino-PEG4-alcohol CAS:86770-74-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW008005100MG
Encompass_Preferred
Medchemexpress, HY-W008005 100mg Amino-PEG4-alcohol CAS:86770-74-3 Amino-PEG4-alcohol is a PEG-based PROTAC linker can be used in the synthesis of PROTACs. Amino-PEG4-alcohol is also a non-cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0654