Learn More
Medchemexpress LLC Amino-PEG4-Boc | 581065-95-4 | 97.0% | C15H31NO6 | 5 G
SDP
Catalog No. 5000357282
Encompass_Preferred
Amino-PEG4-Boc is a PEG-based linker utilized in the synthesis of PROTACs, which are small molecules designed to selectively degrade target proteins by exploiting the ubiquitin-proteasome system. It serves as a key component for connecting different ligands within PROTAC structures.
  • Used in the synthesis of PROTACs.
  • PEG-based linker.
  • Aids in the selective degradation of target proteins.
Catalog No. 50-003-57282 Supplier Medchemexpress LLC Supplier No. HY1401935G
Amino-PEG4-Boc is a PEG-based linker utilized in the synthesis of PROTACs, which are small molecules designed to selectively degrade target proteins by exploiting the ubiquitin-proteasome system. It serves as a key component for connecting different ligands within PROTAC structures.
  • Used in the synthesis of PROTACs.
  • PEG-based linker.
  • Aids in the selective degradation of target proteins.