Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG4-Boc | 581065-95-4 | 97.0% | C15H31NO6 | 500 MG
SDP

Catalog No. 5000357089
Encompass_Preferred

Amino-PEG4-Boc is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. These molecules are designed with two ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein for degradation via the intracellular ubiquitin-proteasome system. It is a colorless to light yellow liquid with a purity of 97.0% and a molecular weight of 321.41.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • Colorless to light yellow liquid appearance
  • Purity of 97.0%
  • Molecular weight of 321.41 g/mol
  • For research use only

Catalog No. 50-003-57089 Supplier Medchemexpress LLC Supplier No. HY140193500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG4-Boc is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. These molecules are designed with two ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein for degradation via the intracellular ubiquitin-proteasome system. It is a colorless to light yellow liquid with a purity of 97.0% and a molecular weight of 321.41.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • Colorless to light yellow liquid appearance
  • Purity of 97.0%
  • Molecular weight of 321.41 g/mol
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.