Learn More
Medchemexpress LLC HY-22335 100mg Medchemexpress, Amino-PEG4-C2-amine CAS:68960-97-4 Purity:>98%
SDP
Catalog No. 501970046
Encompass_Preferred
Medchemexpress, HY-22335 100mg Amino-PEG4-C2-amine CAS:68960-97-4 Amino-PEG4-C2-amine is a PEG-based (4 units) PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0046 Supplier Medchemexpress LLC Supplier No. HY22335100MG
Medchemexpress, HY-22335 100mg Amino-PEG4-C2-amine CAS:68960-97-4 Amino-PEG4-C2-amine is a PEG-based (4 units) PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.