Learn More
Medchemexpress LLC Amino-PEG4-C2-SH hydrochloride | 00-00-0 | 95.0% | C10H24ClNO4S | 10MG
SDP
Catalog No. 5000262014
Encompass_Preferred
Amino-PEG4-C2-SH hydrochloride is a PEG-based, amino-terminated linker bearing a short C2 spacer and a terminal thiol, provided as the hydrochloride salt for use in conjugation chemistry and PROTAC synthesis. It is supplied as a solid with an analytically confirmed purity and available documentation for handling and quality.
  • hydrochloride salt of an amino-terminated PEG4 linker with a C2 thiol handle.
  • suitable for PROTAC linker synthesis and thiol-based conjugation chemistry.
  • provides short PEG spacer to improve solubility and reduce steric hindrance.
  • supplied with certificate of analysis confirming typical purity.
  • available in small milligram pack sizes for research use.
Catalog No. 50-002-62014 Supplier Medchemexpress LLC Supplier No. HY13844610MG
Amino-PEG4-C2-SH hydrochloride is a PEG-based, amino-terminated linker bearing a short C2 spacer and a terminal thiol, provided as the hydrochloride salt for use in conjugation chemistry and PROTAC synthesis. It is supplied as a solid with an analytically confirmed purity and available documentation for handling and quality.
  • hydrochloride salt of an amino-terminated PEG4 linker with a C2 thiol handle.
  • suitable for PROTAC linker synthesis and thiol-based conjugation chemistry.
  • provides short PEG spacer to improve solubility and reduce steric hindrance.
  • supplied with certificate of analysis confirming typical purity.
  • available in small milligram pack sizes for research use.