Learn More
Medchemexpress LLC Amino-PEG4-C2-SH (hydrochloride) | 2762393-21-3 | 95.0% | 289.82 | 25 MG
SDP
Catalog No. 5000350096
Encompass_Preferred
Thiol-PEG4-NH2 hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50096 Supplier Medchemexpress LLC Supplier No. 25MGHY138446
Thiol-PEG4-NH2 hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins