Learn More
Medchemexpress LLC Amino-PEG4-C2-SH hydrochloride | 95.0% | 289.82 g/mol | C10H24ClNO4S | 5 MG
SDP
Catalog No. 5000222426
Encompass_Preferred
Amino-PEG4-C2-SH hydrochloride is a PEG-based PROTAC linker supplied as the hydrochloride salt for research use. The molecule features a PEG4 spacer with a terminal amino group and a terminal thiol, enabling orthogonal conjugation strategies in bifunctional molecule synthesis. It is provided with characterized purity and molecular weight suitable for medicinal chemistry and linker development applications.
  • Hydrochloride salt for improved handling and stability.
  • Terminal amino and thiol groups for orthogonal conjugation.
  • PEG4 spacer provides flexibility and aqueous solubility.
  • Characterized purity for research-grade synthesis.
  • Intended for use in PROTAC and linker chemistry.
Catalog No. 50-002-22426 Supplier Medchemexpress LLC Supplier No. HY1384465MG
Amino-PEG4-C2-SH hydrochloride is a PEG-based PROTAC linker supplied as the hydrochloride salt for research use. The molecule features a PEG4 spacer with a terminal amino group and a terminal thiol, enabling orthogonal conjugation strategies in bifunctional molecule synthesis. It is provided with characterized purity and molecular weight suitable for medicinal chemistry and linker development applications.
  • Hydrochloride salt for improved handling and stability.
  • Terminal amino and thiol groups for orthogonal conjugation.
  • PEG4 spacer provides flexibility and aqueous solubility.
  • Characterized purity for research-grade synthesis.
  • Intended for use in PROTAC and linker chemistry.