Learn More
Medchemexpress LLC Amino-PEG4-CH2COOH | 195071-49-9 | >=98.0% | 251.28 g/mol | C10H21NO6 | 100 MG
SDP
Catalog No. 5000203384
Encompass_Preferred
Amino-PEG4-CH2COOH is a PEG-based bifunctional linker supplied as a purified solid for use in synthetic chemistry. It provides an amino terminus and a terminal carboxylic acid for conjugation and spacer functionality in bifunctional molecules such as PROTACs and antibody-drug conjugates.
  • Bifunctional linker with amino and carboxylic acid termini.
  • Four-unit polyethylene glycol (PEG4) spacer for flexible linkage.
  • High purity suitable for synthesis and research use.
  • Low molecular weight and defined formula for analytical consistency.
  • Available in small-scale research quantities for lab-scale synthesis.
Catalog No. 50-002-03384 Supplier Medchemexpress LLC Supplier No. HY130524100MG
Amino-PEG4-CH2COOH is a PEG-based bifunctional linker supplied as a purified solid for use in synthetic chemistry. It provides an amino terminus and a terminal carboxylic acid for conjugation and spacer functionality in bifunctional molecules such as PROTACs and antibody-drug conjugates.
  • Bifunctional linker with amino and carboxylic acid termini.
  • Four-unit polyethylene glycol (PEG4) spacer for flexible linkage.
  • High purity suitable for synthesis and research use.
  • Low molecular weight and defined formula for analytical consistency.
  • Available in small-scale research quantities for lab-scale synthesis.