Learn More
Medchemexpress LLC Amino-PEG4-CH2COOH | 195071-49-9 | ≥98.0% | 251.28 g/mol | C10H21NO6 | 250 MG
SDP
Catalog No. 5000204123
Encompass_Preferred
Amino-PEG4-CH2COOH is a four-unit polyethylene glycol (PEG4) linker bearing a terminal amino group and a carboxylic acid (-CH2COOH). It is used as a hydrophilic spacer in bioconjugation chemistries such as PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Four-unit PEG spacer providing flexibility and solubility.
  • Terminal amino and carboxylic acid functionalities for straightforward conjugation.
  • High chemical purity suitable for research applications.
  • Molecular weight approximately 251.28 g/mol, formula C10H21NO6.
  • Available in common lab pack sizes, including 250 MG for small-scale synthesis.
Catalog No. 50-002-04123 Supplier Medchemexpress LLC Supplier No. HY130524250MG
Amino-PEG4-CH2COOH is a four-unit polyethylene glycol (PEG4) linker bearing a terminal amino group and a carboxylic acid (-CH2COOH). It is used as a hydrophilic spacer in bioconjugation chemistries such as PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Four-unit PEG spacer providing flexibility and solubility.
  • Terminal amino and carboxylic acid functionalities for straightforward conjugation.
  • High chemical purity suitable for research applications.
  • Molecular weight approximately 251.28 g/mol, formula C10H21NO6.
  • Available in common lab pack sizes, including 250 MG for small-scale synthesis.