Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Amino-Peg4-T-Butyl Ester

Catalog No. 9025351G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-253-51G 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-253-51G Supplier Sigma Aldrich Supplier No. 9025351G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This heterobifunctional, PEGylated crosslinker features an amino group at one end and t-butyl-protected carboxyl group at the other, which can be deprotected with acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. Amino-PEG4-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC™ molecules) for targeted protein degradation.Real-time monitoring of vesicle pH in an endocytic pathway using an EGF-conjugated two-photon probeDevelopment of Substrate-Selective Probes for Affinity Pulldown of Histone DemethylasesA bisphosphonate tweezers and clickable PEGylated PAMAM dendrons for the preparation of functional iron oxide nanoparticles displaying renal and hepatobiliary eliminationPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 2230788-64-2
Molecular Formula C32H49N5O8S -+ xHCl
Linear Formula C32H49N5O8S -+ xHCl
Quantity 50 mg
Synonym tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate; H2N-PEG4-CH2CH2COOtBu; H2N-dPEG(4)-CO-OtBu
Molecular Weight (g/mol) 663.83 (free base basis) g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.