Learn More
Medchemexpress LLC Amino-PEG4-Val-Cit-PAB-MMAE | 1492056-71-9 | 99.9% | 1370.71 | C69H115N11O17 | 1 MG
SDP
Catalog No. 5000202812
Encompass_Preferred
Amino-PEG4-Val-Cit-PAB-MMAE is a cleavable 4-unit PEG antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads such as MMAE to antibodies during ADC synthesis. It contains a Val-Cit dipeptide cleavable spacer and a para-aminobenzyl (PAB) self-immolative spacer with a PEG4 solubilizing linker, providing controlled payload release in target cells.
  • Cleavable Val-Cit peptide spacer enabling enzyme-triggered release
  • PAB self-immolative spacer for efficient payload release
  • PEG4 linker improves solubility and spacer length
  • High reported purity (approximately 99.9%)
  • Soluble in DMSO and compatible with common in vivo formulations
Catalog No. 50-002-02812 Supplier Medchemexpress LLC Supplier No. HY1411541MG
Amino-PEG4-Val-Cit-PAB-MMAE is a cleavable 4-unit PEG antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads such as MMAE to antibodies during ADC synthesis. It contains a Val-Cit dipeptide cleavable spacer and a para-aminobenzyl (PAB) self-immolative spacer with a PEG4 solubilizing linker, providing controlled payload release in target cells.
  • Cleavable Val-Cit peptide spacer enabling enzyme-triggered release
  • PAB self-immolative spacer for efficient payload release
  • PEG4 linker improves solubility and spacer length
  • High reported purity (approximately 99.9%)
  • Soluble in DMSO and compatible with common in vivo formulations