Learn More
Medchemexpress LLC Amino-PEG5-amine | 72236-26-1 | 97.0% | 5 G
SDP
Catalog No. 5000339793
Encompass_Preferred
Amino-PEG5-amine is a PEG-based (5 units) PROTAC linker, appearing as a colorless to light yellow liquid. With a molecular weight of 280.36 and a formula of C12H28N2O5, it is soluble in water (≥ 100 mg/mL). This compound is suitable for research use only.
  • Used in the synthesis of PROTACs
  • Appears as a colorless to light yellow liquid
  • Soluble in water (≥ 100 mg/mL)
  • For research use only
Catalog No. 50-003-39793 Supplier Medchemexpress LLC Supplier No. HY1304475G
Amino-PEG5-amine is a PEG-based (5 units) PROTAC linker, appearing as a colorless to light yellow liquid. With a molecular weight of 280.36 and a formula of C12H28N2O5, it is soluble in water (≥ 100 mg/mL). This compound is suitable for research use only.
  • Used in the synthesis of PROTACs
  • Appears as a colorless to light yellow liquid
  • Soluble in water (≥ 100 mg/mL)
  • For research use only