Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG5-CH2COOH | 141282-35-1 | ≥97.0% | 250 MG
SDP

Catalog No. 5000342248
Encompass_Preferred

Amino-PEG5-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs (proteolysis-targeting chimeras) exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They function by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • PEG-based PROTAC linker
  • Used for synthesizing PROTACs
  • Facilitates selective degradation of target proteins
  • Molecular weight: 295.33
  • Chemical formula: C12H25NO7
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-42248 Supplier Medchemexpress LLC Supplier No. 250MGHY130577
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG5-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs (proteolysis-targeting chimeras) exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They function by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • PEG-based PROTAC linker
  • Used for synthesizing PROTACs
  • Facilitates selective degradation of target proteins
  • Molecular weight: 295.33
  • Chemical formula: C12H25NO7
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.