Learn More
Medchemexpress LLC Amino-PEG5-CH2COOH | 141282-35-1 | ≥97.0% | 500 MG
SDP
Catalog No. 5000340220
Encompass_Preferred
Amino-PEG5-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets the protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Targets PROTAC linkers within the PROTAC pathway
  • Solid appearance
  • White to off-white color
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-40220 Supplier Medchemexpress LLC Supplier No. HY130577500MG
Amino-PEG5-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets the protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Targets PROTAC linkers within the PROTAC pathway
  • Solid appearance
  • White to off-white color
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light