Learn More
Medchemexpress LLC Amino-PEG6-amido-bis-PEG5-N3 | 1147.31 | C48H94N10O21 | 10 MG
SDP
Catalog No. 5000204143
Encompass_Preferred
Amino-PEG6-amido-bis-PEG5-N3 is a cleavable 11-unit polyethylene glycol (PEG) linker containing an amino terminus and an azide functional group. It is supplied as an oil and is intended for use in click-chemistry conjugations and the synthesis of antibody-drug conjugates; for research use only.
  • Cleavable 11-unit PEG linker suitable for bioconjugation.
  • Contains amino and azide functional groups for dual reactivity.
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Supplied as an oil; store sealed away from moisture and light.
  • Available in small laboratory pack sizes for research-scale use.
Catalog No. 50-002-04143 Supplier Medchemexpress LLC Supplier No. HY13095710MG
Amino-PEG6-amido-bis-PEG5-N3 is a cleavable 11-unit polyethylene glycol (PEG) linker containing an amino terminus and an azide functional group. It is supplied as an oil and is intended for use in click-chemistry conjugations and the synthesis of antibody-drug conjugates; for research use only.
  • Cleavable 11-unit PEG linker suitable for bioconjugation.
  • Contains amino and azide functional groups for dual reactivity.
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Supplied as an oil; store sealed away from moisture and light.
  • Available in small laboratory pack sizes for research-scale use.